Assay Details
| Gene Symbol: Fhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033301.4 | NP_001028473.2 | 8 - 9 | 1240 | 98 | |
| NM_001205355.1 | NP_001192284.1 | 8 - 9 | 1311 | 98 | ||
| GenBank mRNA | AK031946.1 | - | 7 - 8 | 713 | 98 | |
| AK145217.1 | - | 8 - 9 | 1231 | 98 | ||
| AK173244.1 | - | 8 - 9 | 1311 | 98 | ||
| BC137632.1 | - | 8 - 9 | 1280 | 8860651 | 98 | |
More Information
Set Membership: |
|

Fhdc1
Genomic Map