Assay Details
| Gene Symbol: Capn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009796.2 | NP_033926.1 | 5 - 6 | 792 | 100 | |
| GenBank mRNA | AB028640.1 | - | 5 - 6 | 792 | 100 | |
| AJ012475.1 | - | 5 - 6 | 605 | 100 | ||
| AK154277.1 | - | 5 - 6 | 878 | 100 | ||
| AK169334.1 | - | 5 - 6 | 837 | 100 | ||
| BC095980.1 | - | 5 - 6 | 809 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn7
Genomic Map