Assay Details
| Gene Symbol: Sh3bp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011894.2 | NP_036024.2 | 2 - 3 | 291 | 59 | |
| GenBank mRNA | AB016835.1 | - | 2 - 3 | 119 | 59 | |
| AK043375.1 | - | 336 | 59 | |||
| AK165033.1 | - | 2 - 3 | 325 | 59 | ||
| BC018237.1 | - | 2 - 3 | 323 | 3482670 | 59 | |
| BC053741.1 | - | 2 - 3 | 291 | 6336511 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3bp5
Genomic Map