Assay Details
| Gene Symbol: Sh3bp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011894.2 | NP_036024.2 | 6 - 7 | 755 | 90 | |
| GenBank mRNA | AB016835.1 | - | 6 - 7 | 583 | 90 | |
| AK043375.1 | - | 800 | 90 | |||
| AK161427.1 | - | 5 - 6 | 737 | 90 | ||
| AK165033.1 | - | 6 - 7 | 789 | 90 | ||
| BC018237.1 | - | 6 - 7 | 787 | 3482670 | 90 | |
| BC053741.1 | - | 6 - 7 | 755 | 6336511 | 90 | |
More Information
Set Membership: |
|

Sh3bp5
Genomic Map