Assay Details
| Gene Symbol: Sema3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025379.1 | NP_001020550.1 | 8 - 9 | 964 | 110 | |
| GenBank mRNA | AB127607.1 | - | 8 - 9 | 964 | 110 | |
| AK129018.1 | - | 8 - 9 | 979 | 110 | ||
| AK137463.1 | - | 8 - 9 | 981 | 110 | ||
| BC115440.2 | - | 8 - 9 | 963 | 40094966 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema3g
Genomic Map