Assay Details
| Gene Symbol: Hacl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019975.3 | NP_064359.2 | 11 - 12 | 1090 | 82 | |
| GenBank mRNA | AJ132139.1 | - | 11 - 12 | 1088 | 82 | |
| AK005505.1 | - | 11 - 12 | 1077 | 82 | ||
| AK041686.1 | - | 11 - 12 | 1086 | 82 | ||
| AK050078.1 | - | 11 - 12 | 1090 | 82 | ||
| AK161488.1 | - | 1070 | 82 | |||
| BC021360.1 | - | 11 - 12 | 1021 | 5011451 | 82 | |
More Information
Set Membership: |
|

Hacl1
Genomic Map