Assay Details
| Gene Symbol: Ncoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033988.2 | NP_001029160.1 | 4 - 5 | 510 | 133 | |
| NM_019744.4 | NP_062718.2 | 4 - 5 | 420 | 133 | ||
| GenBank mRNA | AF159461.1 | - | 4 - 5 | 417 | 133 | |
| AK014479.1 | - | 4 - 5 | 418 | 133 | ||
| AK032951.1 | - | 2 - 3 | 176 | 133 | ||
| AK129020.1 | - | 4 - 5 | 468 | 133 | ||
| AK132981.1 | - | 133 | ||||
| AK143965.1 | - | 4 - 5 | 415 | 133 | ||
| AK145070.1 | - | 4 - 5 | 471 | 133 | ||
| AK145945.1 | - | 4 - 5 | 463 | 133 | ||
| AK146384.1 | - | 4 - 5 | 421 | 133 | ||
| AK146498.1 | - | 4 - 5 | 460 | 133 | ||
| AK146598.1 | - | 4 - 5 | 444 | 133 | ||
| AK146646.1 | - | 4 - 5 | 415 | 133 | ||
| AK146649.1 | - | 4 - 5 | 417 | 133 | ||
| AK147036.1 | - | 4 - 5 | 418 | 133 | ||
| AK147125.1 | - | 4 - 5 | 462 | 133 | ||
| BC031528.1 | - | 4 - 5 | 461 | 4008986 | 133 | |
| BC085086.1 | - | 4 - 5 | 406 | 6335192 | 133 | |
| CF161810.1 | - | 4 - 5 | 505 | 133 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ncoa4
Genomic Map