Assay Details
| Gene Symbol: Eaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028932.4 | NP_083208.1 | 4 - 5 | 720 | 69 | |
| GenBank mRNA | AK016628.1 | - | 1401 | 69 | ||
| AK036661.1 | - | 4 - 5 | 692 | 69 | ||
| AK044181.1 | - | 4 - 5 | 721 | 69 | ||
| AK044737.1 | - | 4 - 5 | 657 | 69 | ||
| BC079658.1 | - | 4 - 5 | 697 | 6853920 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eaf1
Genomic Map