Assay Details
| Gene Symbol: Galnt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030166.3 | NP_084442.1 | 2 - 3 | 1092 | 107 | |
| GenBank mRNA | AB175682.1 | - | 2 - 3 | 702 | 107 | |
| AK019470.1 | - | 2 - 3 | 1207 | 107 | ||
| AK030347.1 | - | 2 - 3 | 344 | 107 | ||
| AK050075.1 | - | 2 - 3 | 590 | 108 | ||
| AK163883.1 | - | 2 - 3 | 968 | 107 | ||
| BC133711.1 | - | 2 - 3 | 1092 | 40143107 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt15
Genomic Map