Assay Details
| Gene Symbol: Morc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045529.3 | NP_001038994.2 | 14 - 15 | 1821 | 87 | |
| GenBank mRNA | AK132226.1 | - | 13 - 14 | 1781 | 87 | |
| AK172896.1 | - | 14 - 15 | 1821 | 87 | ||
| BC026506.1 | - | 1 - 2 | 174 | 87 | ||
| BC029284.1 | - | 1 - 2 | 51 | 87 | ||
| BC098072.1 | - | 14 - 15 | 1662 | 30109159 | 87 | |
| BC145705.1 | - | 14 - 15 | 1770 | 40131008 | 87 | |
More Information
Set Membership: |
|

Morc3
Genomic Map