Assay Details
| Gene Symbol: Morc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045529.3 | NP_001038994.2 | 15 - 16 | 2708 | 96 | |
| GenBank mRNA | AK168211.1 | - | 6 - 7 | 1394 | 95 | |
| AK172896.1 | - | 15 - 16 | 2708 | 96 | ||
| BC026506.1 | - | 2 - 3 | 1061 | 96 | ||
| BC029284.1 | - | 2 - 3 | 938 | 96 | ||
| BC098072.1 | - | 15 - 16 | 2549 | 30109159 | 96 | |
| BC145705.1 | - | 15 - 16 | 2657 | 40131008 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Morc3
Genomic Map