Assay Details
| Gene Symbol: Morc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045529.3 | NP_001038994.2 | 3 - 4 | 433 | 86 | |
| GenBank mRNA | AK132226.1 | - | 3 - 4 | 289 | 86 | |
| AK172896.1 | - | 3 - 4 | 433 | 86 | ||
| BC098072.1 | - | 3 - 4 | 274 | 30109159 | 86 | |
| BC145705.1 | - | 3 - 4 | 382 | 40131008 | 86 | |
More Information
Set Membership: |
|

Morc3
Genomic Map