Assay Details
| Gene Symbol: Morc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045529.3 | NP_001038994.2 | 10 - 11 | 1400 | 82 | |
| GenBank mRNA | AK132226.1 | - | 10 - 11 | 1256 | 82 | |
| AK168211.1 | - | 1 - 2 | 87 | 82 | ||
| AK172896.1 | - | 10 - 11 | 1400 | 82 | ||
| BC098072.1 | - | 10 - 11 | 1241 | 30109159 | 82 | |
| BC145705.1 | - | 10 - 11 | 1349 | 40131008 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Morc3
Genomic Map