Assay Details
| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028881.2 | NP_083157.1 | 5 - 6 | 631 | 87 | |
| GenBank mRNA | AK156192.1 | - | 5 - 6 | 610 | 87 | |
| AK172084.1 | - | 5 - 6 | 589 | 87 | ||
| BC023831.1 | - | 5 - 6 | 626 | 5345301 | 87 | |
| BC032183.1 | - | 5 - 6 | 636 | 5310152 | 87 | |
| BC033300.1 | - | 5 - 6 | 626 | 4166411 | 87 | |
| BU612513.1 | - | 5 - 6 | 572 | 87 | ||
| CV783724.1 | - | 2 - 3 | 135 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crtc2
Genomic Map