Assay Details
| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028881.2 | NP_083157.1 | 8 - 9 | 831 | 76 | |
| GenBank mRNA | AK014553.1 | - | 3 - 4 | 1202 | 76 | |
| AK079382.1 | - | 6 - 7 | 1579 | 76 | ||
| AK156192.1 | - | 8 - 9 | 810 | 76 | ||
| AK169944.1 | - | 4 - 5 | 480 | 76 | ||
| AK172084.1 | - | 8 - 9 | 789 | 76 | ||
| BC023091.1 | - | 4 - 5 | 421 | 4023064 | 76 | |
| BC023831.1 | - | 8 - 9 | 826 | 5345301 | 76 | |
| BC032183.1 | - | 8 - 9 | 836 | 5310152 | 76 | |
| BC033300.1 | - | 8 - 9 | 826 | 4166411 | 76 | |
| CV783724.1 | - | 5 - 6 | 335 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crtc2
Genomic Map