Assay Details
| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027315.4 | NP_081591.3 | 8 - 9 | 1066 | 91 | |
| GenBank mRNA | AK009324.1 | - | 7 - 8 | 648 | 91 | |
| AK146707.1 | - | 8 - 9 | 879 | 91 | ||
| AY112699.1 | - | 8 - 9 | 1059 | 91 | ||
| AY945937.1 | - | 8 - 9 | 963 | 91 | ||
| BC030044.1 | - | 5 - 6 | 391 | 91 | ||
| BC051487.1 | - | 1 - 2 | 63 | 91 | ||
| BC060966.1 | - | 6 - 7 | 459 | 91 | ||
| BC082275.1 | - | 8 - 9 | 889 | 6310111 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2q1
Genomic Map