Assay Details
| Gene Symbol: Npr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008727.5 | NP_032753.5 | 13 - 14 | 2449 | 134 | |
| GenBank mRNA | AK135008.1 | - | 4 - 5 | 392 | 134 | |
| BC110659.1 | - | 13 - 14 | 2329 | 30847317 | 134 | |
| BC139767.1 | - | 13 - 14 | 2308 | 134 | ||
| J05504.1 | - | 14 - 15 | 2433 | 134 | ||
| L31932.1 | - | 13 - 14 | 2097 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npr1
Genomic Map