Assay Details
| Gene Symbol: Npr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008727.5 | NP_032753.5 | 19 - 20 | 3282 | 108 | |
| GenBank mRNA | AK135008.1 | - | 10 - 11 | 1225 | 108 | |
| BC110659.1 | - | 19 - 20 | 3162 | 30847317 | 108 | |
| BC139767.1 | - | 19 - 20 | 3141 | 108 | ||
| L31932.1 | - | 19 - 20 | 2930 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npr1
Genomic Map