Assay Details
| Gene Symbol: Ncapg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019438.1 | NP_062311.1 | 14 - 15 | 2200 | 80 | |
| GenBank mRNA | AK136254.1 | - | 9 - 10 | 1186 | 80 | |
| BC137834.1 | - | 7 - 8 | 1418 | 8860853 | 80 | |
| BC144960.1 | - | 7 - 8 | 1418 | 9053508 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncapg
Genomic Map