Assay Details
| Gene Symbol: Ncapg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019438.1 | NP_062311.1 | 17 - 18 | 2717 | 91 | |
| GenBank mRNA | AJ237585.1 | - | 11 - 12 | 3100 | 91 | |
| AK136254.1 | - | 12 - 13 | 1703 | 91 | ||
| BC137834.1 | - | 10 - 11 | 1935 | 8860853 | 91 | |
| BC144960.1 | - | 10 - 11 | 1935 | 9053508 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncapg
Genomic Map