Assay Details
| Gene Symbol: Ctso | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177662.2 | NP_808330.1 | 3 - 4 | 406 | 72 | |
| GenBank mRNA | AK034490.1 | - | 3 - 4 | 406 | 72 | |
| AK049470.1 | - | 3 - 4 | 379 | 72 | ||
| AK143683.1 | - | 3 - 4 | 422 | 72 | ||
| AK165930.1 | - | 3 - 4 | 368 | 72 | ||
| AK166103.1 | - | 3 - 4 | 375 | 72 | ||
| BC044664.1 | - | 3 - 4 | 298 | 72 | ||
| BC098219.1 | - | 3 - 4 | 380 | 1066724 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctso
Genomic Map