Assay Details
| Gene Symbol: Med28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025895.4 | NP_080171.1 | 2 - 3 | 323 | 103 | |
| GenBank mRNA | AF318060.1 | - | 2 - 3 | 247 | 103 | |
| AK003854.1 | - | 2 - 3 | 236 | 103 | ||
| AK005681.1 | - | 2 - 3 | 232 | 103 | ||
| AK007855.1 | - | 2 - 3 | 252 | 103 | ||
| AK154249.1 | - | 2 - 3 | 249 | 103 | ||
| BC024941.1 | - | 2 - 3 | 266 | 4017146 | 103 | |
| BY725533.1 | - | 2 - 3 | 303 | 103 | ||
| DV653139.1 | - | 2 - 3 | 323 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Med28
Genomic Map