Assay Details
| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178394.4 | NP_848481.2 | 11 - 12 | 1897 | 67 | |
| GenBank mRNA | AK077298.1 | - | 11 - 12 | 1839 | 67 | |
| BC138646.1 | - | 10 - 11 | 1749 | 8861666 | 67 | |
| BC138649.1 | - | 10 - 11 | 1749 | 8861669 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jakmip1
Genomic Map