Assay Details
| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178394.4 | NP_848481.2 | 4 - 5 | 1086 | 60 | |
| GenBank mRNA | AK017977.1 | - | 4 - 5 | 1041 | 60 | |
| AK077298.1 | - | 4 - 5 | 1028 | 60 | ||
| BC138646.1 | - | 3 - 4 | 938 | 8861666 | 60 | |
| BC138649.1 | - | 3 - 4 | 938 | 8861669 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jakmip1
Genomic Map