Assay Details
| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178394.4 | NP_848481.2 | 7 - 8 | 1497 | 74 | |
| GenBank mRNA | AK077298.1 | - | 7 - 8 | 1439 | 74 | |
| BC138646.1 | - | 6 - 7 | 1349 | 8861666 | 74 | |
| BC138649.1 | - | 6 - 7 | 1349 | 8861669 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jakmip1
Genomic Map