Assay Details
| Gene Symbol: Sox18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009236.2 | NP_033262.2 | 1 - 2 | 457 | 74 | |
| GenBank mRNA | AK144281.1 | - | 1 - 2 | 458 | 74 | |
| BC006612.1 | - | 1 - 2 | 457 | 3967084 | 74 | |
| L29086.1 | - | 1 - 2 | 110 | 74 | ||
| L35032.1 | - | 1 - 2 | 426 | 74 | ||
More Information
Set Membership: |
|

Sox18
Genomic Map