Assay Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133701.2 | NP_598462.1 | 15 - 16 | 2173 | 112 | |
| GenBank mRNA | AK081998.1 | - | 15 - 16 | 2108 | 112 | |
| AK145461.1 | - | 15 - 16 | 2176 | 112 | ||
| AK166890.1 | - | 15 - 16 | 2125 | 112 | ||
| BC005801.1 | - | 5 - 6 | 689 | 112 | ||
| BC014869.1 | - | 15 - 16 | 2100 | 3602158 | 112 | |
| BC023691.1 | - | 15 - 16 | 2102 | 5344332 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf6
Genomic Map