Assay Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133701.2 | NP_598462.1 | 16 - 17 | 2344 | 74 | |
| GenBank mRNA | AK004462.1 | - | 1 - 2 | 44 | 74 | |
| AK081998.1 | - | 16 - 17 | 2279 | 74 | ||
| AK145461.1 | - | 16 - 17 | 2347 | 74 | ||
| AK166890.1 | - | 16 - 17 | 2296 | 74 | ||
| BC005801.1 | - | 6 - 7 | 860 | 74 | ||
| BC014869.1 | - | 16 - 17 | 2271 | 3602158 | 74 | |
| BC023691.1 | - | 16 - 17 | 2273 | 5344332 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf6
Genomic Map