Assay Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133701.2 | NP_598462.1 | 2 - 3 | 378 | 74 | |
| GenBank mRNA | AK011639.1 | - | 2 - 3 | 320 | 74 | |
| AK081998.1 | - | 2 - 3 | 313 | 74 | ||
| AK145461.1 | - | 2 - 3 | 381 | 74 | ||
| AK166890.1 | - | 2 - 3 | 330 | 74 | ||
| BC014869.1 | - | 2 - 3 | 305 | 3602158 | 74 | |
| BC023691.1 | - | 2 - 3 | 307 | 5344332 | 74 | |
| BC025030.1 | - | 2 - 3 | 324 | 4947416 | 74 | |
| BY735790.1 | - | 2 - 3 | 378 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf6
Genomic Map