Assay Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133701.2 | NP_598462.1 | 7 - 8 | 1014 | 117 | |
| GenBank mRNA | AK081998.1 | - | 7 - 8 | 949 | 117 | |
| AK145461.1 | - | 7 - 8 | 1017 | 117 | ||
| AK166890.1 | - | 7 - 8 | 966 | 117 | ||
| BC014869.1 | - | 7 - 8 | 941 | 3602158 | 117 | |
| BC023691.1 | - | 7 - 8 | 943 | 5344332 | 117 | |
| BC025030.1 | - | 7 - 8 | 960 | 4947416 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf6
Genomic Map