Assay Details
| Gene Symbol: Srms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011481.3 | NP_035611.3 | 4 - 5 | 1231 | 73 | |
| GenBank mRNA | AK049582.1 | - | 4 - 5 | 492 | 73 | |
| BC120633.1 | - | 4 - 5 | 1210 | 40129556 | 73 | |
| BC125325.1 | - | 4 - 5 | 1207 | 40129840 | 73 | |
| D26186.1 | - | 4 - 5 | 1077 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srms
Genomic Map