Assay Details
| Gene Symbol: Srms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011481.3 | NP_035611.3 | 5 - 6 | 1387 | 87 | |
| GenBank mRNA | AK049582.1 | - | 5 - 6 | 648 | 87 | |
| BC120633.1 | - | 5 - 6 | 1366 | 40129556 | 87 | |
| BC125325.1 | - | 5 - 6 | 1363 | 40129840 | 87 | |
| D26186.1 | - | 5 - 6 | 1233 | 87 | ||
| D49427.1 | - | 5 - 6 | 1401 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srms
Genomic Map