Assay Details
| Gene Symbol: Helz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183162.2 | NP_898985.2 | 8 - 9 | 3461 | 55 | |
| GenBank mRNA | AB201714.1 | - | 7 - 8 | 3365 | 55 | |
| AK157942.1 | - | 8 - 9 | 3460 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Helz2
Genomic Map