Assay Details
| Gene Symbol: Slc4a1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009206.2 | NP_033232.2 | 8 - 9 | 1631 | 84 | |
| GenBank mRNA | AF035526.1 | - | 9 - 10 | 1514 | 84 | |
| BC090964.1 | - | 8 - 9 | 1183 | 30625511 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a1ap
Genomic Map