Assay Details
| Gene Symbol: Supt7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028150.2 | NP_082426.1 | 2 - 3 | 754 | 64 | |
| GenBank mRNA | AK012134.1 | - | 2 - 3 | 700 | 64 | |
| AK047063.1 | - | 2 - 3 | 724 | 64 | ||
| AK144989.1 | - | 2 - 3 | 626 | 64 | ||
| BC031447.1 | - | 2 - 3 | 677 | 4502314 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt7l
Genomic Map