Assay Details
| Gene Symbol: Agbl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048192.2 | NP_001041657.1 | 16 - 17 | 2744 | 130 | |
| NR_122116.1 | 15 - 16 | 2819 | 130 | |||
| NR_122117.1 | 15 - 16 | 2728 | 130 | |||
| GenBank mRNA | AK035680.1 | - | 13 - 14 | 2229 | 130 | |
| AK083656.1 | - | 14 - 15 | 2916 | 130 | ||
| BC057349.1 | - | 15 - 16 | 2753 | 6813901 | 130 | |
| DQ867034.1 | - | 15 - 16 | 2748 | 130 | ||
| DQ867035.1 | - | 15 - 16 | 2657 | 130 | ||
| DQ867036.1 | - | 16 - 17 | 2744 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agbl5
Genomic Map