| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400018954.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018954.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018954.mm.1 |
TC0400002295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018953.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018953.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018953.mm.1 |
TC0400002295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018955.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018955.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018955.mm.1 |
TC0400002295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018958.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018958.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018958.mm.1 |
TC0400002295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018952.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018952.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018952.mm.1 |
TC0400002295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018956.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018956.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018956.mm.1 |
TC0400002295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018959.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018959.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018959.mm.1 |
TC0400002295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018957.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018957.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018957.mm.1 |
TC0400002295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018960.mm.1 |
NM_008899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018960.mm.1 |
NM_008899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018960.mm.1 |
TC0400002295.mm.1 |
MTA TC |