Assay Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029162.2 | NP_083438.1 | 3 - 4 | 1350 | 84 | |
| GenBank mRNA | AK043044.1 | - | 3 - 4 | 1345 | 84 | |
| AK140546.1 | - | 3 - 4 | 1345 | 84 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK148068.1 | - | 1356 | Chr. - - [-] on Build | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb49
Genomic Map