Assay Details
| Gene Symbol: Rarres1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164763.1 | NP_001158235.1 | 2 - 3 | 428 | 83 | |
| GenBank mRNA | AK077393.1 | - | 2 - 3 | 428 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P401184 | AK077393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P401184 | NM_001164763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P401184 | AK077393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P401184 | NM_001164763.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143808 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157921 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145318 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P401184 | AK077393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P401184 | NM_001164763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P401184 | AK077393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P401184 | NM_001164763.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157921 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145318 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143808 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00663640 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00663656 | - | - |
| Whole Mouse Genome G4122A | A_51_P401184 | AK077393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P401184 | NM_001164763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P401184 | AK077393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P401184 | NM_001164763.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143808 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157921 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02411020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02635477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02446705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02411023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02764831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02522373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02764820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02719539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02666528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02635481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02635474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02509253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02411016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02653544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02328239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02635475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02509249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02374237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02653553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02392626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03104804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02522369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03020144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02719562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02411019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02522368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22210477 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1438055_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10498584 | - | - |
| Mouse Genome 430 2.0 Array | 1438055_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438055_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109222_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109222_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300015974.mm.1 | NM_001164763.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015974.mm.1 | NM_001164763.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015974.mm.1 | TC0300002114.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438055_at | - | - |
| Mouse Expression Set 430 | 1438055_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438055_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438055_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 445847 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5860390 | - | - |
| MouseWG-6 v2 Expression BeadChip | 770139 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605971 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664202 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2605971 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664202 | - | - |

Rarres1
Genomic Map