Assay Details
| Gene Symbol: Rarres1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164763.1 | NP_001158235.1 | 3 - 4 | 613 | 88 | |
| GenBank mRNA | AK077393.1 | - | 3 - 4 | 613 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rarres1
Genomic Map