Assay Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025813.3 | NP_080089.1 | 10 - 11 | 967 | 68 | |
| GenBank mRNA | AK004586.1 | - | 10 - 11 | 968 | 68 | |
| AK032688.1 | - | 10 - 11 | 935 | 68 | ||
| AK075859.1 | - | 10 - 11 | 997 | 68 | ||
| AK135235.1 | - | 10 - 11 | 968 | 68 | ||
| AK136108.1 | - | 10 - 11 | 959 | 68 | ||
| AK152444.1 | - | 10 - 11 | 989 | 68 | ||
| AK159554.1 | - | 10 - 11 | 995 | 68 | ||
| AK166645.1 | - | 4 - 5 | 359 | 68 | ||
| AK208838.1 | - | 2 - 3 | 84 | 68 | ||
| BC024891.1 | - | 10 - 11 | 949 | 3992247 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd1
Genomic Map