Assay Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025813.3 | NP_080089.1 | 3 - 4 | 379 | 88 | |
| GenBank mRNA | AK004586.1 | - | 3 - 4 | 380 | 88 | |
| AK032688.1 | - | 3 - 4 | 348 | 88 | ||
| AK075859.1 | - | 3 - 4 | 409 | 88 | ||
| AK135235.1 | - | 3 - 4 | 380 | 88 | ||
| AK136108.1 | - | 3 - 4 | 371 | 88 | ||
| AK152444.1 | - | 3 - 4 | 401 | 88 | ||
| AK159554.1 | - | 3 - 4 | 407 | 88 | ||
More Information
Set Membership: |
|

Mfsd1
Genomic Map