Assay Details
| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027276.2 | NP_081552.2 | 10 - 11 | 1017 | 104 | |
| GenBank mRNA | AK147938.1 | - | 10 - 11 | 1032 | 104 | |
| AK167965.1 | - | 10 - 11 | 1032 | 104 | ||
| AK168978.1 | - | 10 - 11 | 1004 | 104 | ||
| AK179999.1 | - | 361 | 104 | |||
| BC026606.1 | - | 10 - 11 | 1017 | 4976424 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc16
Genomic Map