Assay Details
| Gene Symbol: Tfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291765.1 | NP_001278694.1 | 2 - 3 | 134 | 70 | |
| NM_009361.3 | NP_033387.1 | 2 - 3 | 195 | 70 | ||
| GenBank mRNA | AK019997.1 | - | 2 - 3 | 135 | 70 | |
| AK030820.1 | - | 2 - 3 | 196 | 70 | ||
| BC058633.1 | - | 2 - 3 | 178 | 70 | ||
| BC132567.1 | - | 1 - 2 | 40 | 40130844 | 70 | |
| BC132569.1 | - | 1 - 2 | 40 | 40130846 | 70 | |
| BC145344.1 | - | 1 - 2 | 40 | 9053900 | 70 | |
| X72310.1 | - | 1 - 2 | 69 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019997.1, AK030820.1, BC132567.1, BC132569.1, BC145344.1, NM_001291765.1, NM_009361.3, X72310.1 transcripts.
Set Membership: |
|

Tfdp1
Genomic Map