Assay Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146207.2 | NP_666319.2 | 7 - 8 | 800 | 64 | |
| GenBank mRNA | AK144802.1 | - | 7 - 8 | 782 | 64 | |
| AK150237.1 | - | 7 - 8 | 779 | 64 | ||
| AK168215.1 | - | 7 - 8 | 782 | 64 | ||
| AK170722.1 | - | 7 - 8 | 782 | 64 | ||
| BC010211.1 | - | 7 - 8 | 778 | 3969515 | 64 | |
| BC024113.1 | - | 3 - 4 | 272 | 5097944 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cul4a
Genomic Map