Assay Details
| Gene Symbol: Ripk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023663.6 | NP_076152.2 | 4 - 5 | 717 | 98 | |
| GenBank mRNA | AF302127.3 | - | 4 - 5 | 720 | 98 | |
| AK145203.1 | - | 4 - 5 | 699 | 98 | ||
| BC057871.1 | - | 4 - 5 | 680 | 5294197 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ripk4
Genomic Map