Assay Details
| Gene Symbol: B3galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020026.4 | NP_064410.1 | 2 - 3 | 231 | 69 | |
| GenBank mRNA | AK088407.1 | - | 2 - 3 | 134 | 69 | |
| BY278563.1 | - | 2 - 3 | 153 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK088407.1, NM_020026.4 transcripts.
Set Membership: |
|

B3galnt1
Genomic Map