Assay Details
| Gene Symbol: Ptpn20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008978.2 | NP_033004.1 | 7 - 8 | 725 | 98 | |
| GenBank mRNA | AK017003.1 | - | 7 - 8 | 788 | 98 | |
| AK029493.1 | - | 7 - 8 | 723 | 98 | ||
| D64141.1 | - | 6 - 7 | 724 | 98 | ||
More Information
Set Membership: |
|

Ptpn20
Genomic Map