Assay Details
| Gene Symbol: Anxa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281845.1 | NP_001268774.1 | 5 - 6 | 484 | 84 | |
| NM_013473.4 | NP_038501.2 | 5 - 6 | 484 | 84 | ||
| GenBank mRNA | AJ002390.1 | - | 5 - 6 | 460 | 84 | |
| AK009085.1 | - | 5 - 6 | 446 | 84 | ||
| AK075870.1 | - | 5 - 6 | 418 | 84 | ||
| BC013271.1 | - | 5 - 6 | 449 | 4013266 | 84 | |
| BC030407.1 | - | 5 - 6 | 497 | 5322310 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anxa8
Genomic Map