Assay Details
| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145711.4 | NP_663757.3 | 5 - 6 | 1147 | 111 | |
| GenBank mRNA | AF472514.1 | - | 5 - 6 | 943 | 111 | |
| AK029694.1 | - | 5 - 6 | 944 | 111 | ||
| AK038671.1 | - | 5 - 6 | 1150 | 111 | ||
| AK133397.1 | - | 5 - 6 | 944 | 111 | ||
| AK161578.1 | - | 5 - 6 | 933 | 111 | ||
| BC038873.1 | - | 1 - 2 | 163 | 111 | ||
| BC080732.1 | - | 5 - 6 | 1064 | 6518356 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tox
Genomic Map